2-Chloro-4-iodopyridine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
C2089-5G
C2089-1G
TCIAC2089-5GEA
710
SEK
TCIAC2089-5G
TCIAC2089-1G
2-Chloro-4-iodopyridine ≥98.0%
2-Chloro-4-iodopyridine
Formel:
C₅H₃ClIN MW: 239,44 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD01861983 CAS nummer: 153034-86-7 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...