3-Chlorobenzophenone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
m-Chlorobenzophenone
C1775-25G
C1775-500G
TCIAC1775-25GEA
565
SEK
TCIAC1775-25G
TCIAC1775-500G
3-Chlorobenzophenone ≥98.0%
3-Chlorobenzophenone
Formel:
C₁₃H₉ClO MW: 216,67 g/mol Kokpunkt: 332°C Smältpunkt: 83°C Storage Temperature: Omgivande |
MDL Number:
MFCD00009816 CAS nummer: 1016-78-0 EINECS: 213-809-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...