2,6-Dichloropyrazine ≥99.0%

Leverantör: TCI EUROPE
Varning

D1867-25G D1867-5G
TCIAD1867-25GEA 2900 SEK
TCIAD1867-25G TCIAD1867-5G
2,6-Dichloropyrazine ≥99.0%
2,6-Dichloropyrazine
Formel: C₄H₂Cl₂N₂
MW: 148,98 g/mol
Kokpunkt: 122°C
Smältpunkt: 57…58 °C
Densitet: 1,713 g/cm³ (20 °C)
MDL Number: MFCD00006125
CAS nummer: 4774-14-5
EINECS: 225-316-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8338b23f3d14fb90fc280984122d54fc610acf8b&vid=8338b23f3d14fb90fc280984122d54fc610acf8b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 53.0 to 56.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR