5-Chloroindole ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 5-Chloro-1H-indole
C1107-25G C1107-5G
TCIAC1107-25GEA 1425 SEK
TCIAC1107-25G TCIAC1107-5G
5-Chloroindole ≥98.0%
5-Chloroindole
Formel: C₈H₆ClN
MW: 151,6 g/mol
Kokpunkt: 130°C
Smältpunkt: 71…72 °C
MDL Number: MFCD00005672
CAS nummer: 17422-32-1
EINECS: 241-448-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=060b2e507efab53205d8b2076e08a722d2bc77ed&vid=060b2e507efab53205d8b2076e08a722d2bc77ed&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C
Solubility in Methanol almost transparency