4,4 '-DDE ≥99.0%
Leverantör: TCI
Fara
B0133-1G B0133-5G
TCIAB0133-1GEA 855 SEK
TCIAB0133-1G TCIAB0133-5G
4,4 '-DDE ≥99.0%
4,4 '-DDE
Formel: C₁₄H₈Cl₄
Kokpunkt: 316,5 °C (1013 hPa)
Smältpunkt: 88…90 °C
Densitet: 1,506 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000837
CAS nummer: 72-55-9
EINECS: 200-784-6
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47568d690988bb3f575666852e1b4586d13c1a8a&vid=47568d690988bb3f575666852e1b4586d13c1a8a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 88.0 to 90.0 deg-C
Solubility in hot Methanol almost transparency