α-Chloro-4-ethyltoulene ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1-(Chloromethyl)-4-ethylbenzene, α-Chloro-p-ethyltoulene, 4-Ethylbenzyl chloride
E0738-25G
E0738-5G
TCIAE0738-25GEA
2150
SEK
TCIAE0738-25G
TCIAE0738-5G
α-Chloro-4-ethyltoulene ≥98.0%
α-Chloro-4-ethyltoulene
Formel:
C₉H₁₁Cl MW: 154,64 g/mol Kokpunkt: 111°C |
MDL Number:
MFCD00039357 CAS nummer: 1467-05-6 UN: 3265 ADR: 8,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...