3-Cyanobenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Formylbenzonitrile
C0429-25G
C0429-5G
TCIAC0429-25GEA
3100
SEK
TCIAC0429-25G
TCIAC0429-5G
3-Cyanobenzaldehyde ≥98.0%
3-Cyanobenzaldehyde
Formel:
C₈H₅NO MW: 131,13 g/mol Kokpunkt: 210°C Smältpunkt: 77…79 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00003344 CAS nummer: 24964-64-5 EINECS: 246-549-1 ADR: 6.1,II |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 80.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...