3-Bromophenyl isocyanate ≥98.0%
Leverantör: TCI
Fara
I0553-25G I0553-5G
TCIAI0553-25GEA 5850 SEK
TCIAI0553-25G TCIAI0553-5G
3-Bromophenyl isocyanate ≥98.0%
3-Bromophenyl isocyanate
Formel: C₇H₄BrNO
Kokpunkt: 220°C
Densitet: 1,586 g/cm³ (20 °C)
Flampunkt: 108°C
Storage Temperature: Kylskåp
MDL Number: MFCD00002014
CAS nummer: 23138-55-8
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83f2a1e258a68441fb925392d9109dc754b8ef7c&vid=83f2a1e258a68441fb925392d9109dc754b8ef7c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid to slightly cloudy liquid
Purity (GC) min. 98.0 %