3-Bromobenzoyl chloride ≥98.0%
Leverantör: TCI
Fara
Synonyms: m-Bromobenzoyl chloride, 3-Bromobenzoylchloride
B0557-25G B0557-5G
TCIAB0557-25GEA 1100 SEK
TCIAB0557-25G TCIAB0557-5G
3-Bromobenzoyl chloride ≥98.0%
3-Bromobenzoyl chloride
Formel: C₇H₄BrClO
Kokpunkt: 240°C
Densitet: 1,66 g/cm³ (20 °C)
Flampunkt: 107°C
Storage Temperature: Omgivande
MDL Number: MFCD00000669
CAS nummer: 1711-09-7
EINECS: 216-978-9
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61ecd6c5ebc6ab7f50ea351d29c6941221496a9c&vid=61ecd6c5ebc6ab7f50ea351d29c6941221496a9c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 98.0 %