Fenantren ≥97.0%

Leverantör: TCI
Varning

P0079-25G P0079-500G
TCIAP0079-25GEA 630 SEK
TCIAP0079-25G TCIAP0079-500G
Fenantren ≥97.0%
Fenantren
Formel: C₁₄H₁₀
Kokpunkt: 340°C
Smältpunkt: 97…99 °C
Densitet: 1,19 g/cm³ (20 °C)
Flampunkt: 153 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001168
CAS nummer: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9,III
Merck Index: 14,07212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ccd4a98a1605ed136fdf8d3f7034682d763b43b&vid=2ccd4a98a1605ed136fdf8d3f7034682d763b43b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 98.0 to 102.0 deg-C
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR