Diphenylsilane ≥98.0%

Leverantör: TCI EUROPE
Varning

D2820-25G D2820-5G
TCIAD2820-25GEA 2525 SEK
TCIAD2820-25G TCIAD2820-5G
Diphenylsilane ≥98.0%
Diphenylsilane
Formel: C₁₂H₁₂Si
MW: 184,31 g/mol
Kokpunkt: 97°C
Densitet: 0,9962 g/cm³ (20 °C)
Flampunkt: 98°C
Storage Temperature: Omgivande
MDL Number: MFCD00003002
CAS nummer: 775-12-2
EINECS: 212-271-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b13b907f2ee6ffbdb0e97d6bb9cc2aa3786ea463&vid=b13b907f2ee6ffbdb0e97d6bb9cc2aa3786ea463&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR