Diphenylsilane ≥97.0%
Leverantör: TCI EUROPE
Varning
D2406-25G D2406-5G
TCIAD2406-25GEA 2225 SEK
TCIAD2406-25G TCIAD2406-5G
Diphenylsilane ≥97.0%
Diphenylsilane
Formel: C₁₂H₁₂Si
MW: 184,31 g/mol
Kokpunkt: 127°C
Densitet: 0,9962 g/cm³ (20 °C)
Flampunkt: 98°C
Storage Temperature: Omgivande
MDL Number: MFCD00003002
CAS nummer: 775-12-2
EINECS: 212-271-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea1204ae0fc5fd1739c10204a69fd067d024c3ee&vid=ea1204ae0fc5fd1739c10204a69fd067d024c3ee&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %