1,1,3,3-Tetramethylguanidine ≥99.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: N,N,N',N'-Tetramethylguanidine

T0148-25ML T0148-500ML T0148-100ML
TCIAT0148-25MLEA 215 SEK
TCIAT0148-25ML TCIAT0148-500ML TCIAT0148-100ML
1,1,3,3-Tetramethylguanidine ≥99.0% (by GC, titration analysis)
1,1,3,3-Tetramethylguanidine
Formel: C₅H₁₃N₃
MW: 115,18 g/mol
Kokpunkt: 163°C
Smältpunkt: –30 °C
Densitet: 0,92 g/cm³ (20 °C)
Flampunkt: 60°C
Storage Temperature: Omgivande
MDL Number: MFCD00008323
CAS nummer: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=826b8c08510a79c2ccc425c9719ef2cec4755532&vid=826b8c08510a79c2ccc425c9719ef2cec4755532&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR