2,5-Dimethylfuran ≥98.0%
Leverantör: TCI
|
Fara
|
D0725-25ML
D0725-100ML
TCIAD0725-25MLEA
335
SEK
TCIAD0725-25ML
TCIAD0725-100ML
2,5-Dimethylfuran ≥98.0%
2,5-Dimethylfuran
Formel:
C₆H₈O Kokpunkt: 92 °C (1013 hPa) Smältpunkt: –62 °C Densitet: 0,903 g/cm³ (20 °C) Flampunkt: 16°C Storage Temperature: Omgivande |
MDL Number:
MFCD00003250 CAS nummer: 625-86-5 EINECS: 210-914-3 UN: 1993 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...