Fluoren ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 9H-Fluorene, Diphenylenmethane
F0017-25G F0017-500G
TCIAF0017-25GEA 215 SEK
TCIAF0017-25G TCIAF0017-500G
Fluoren ≥95.0%
Fluoren
Formel: C₁₃H₁₀
MW: 166,22 g/mol
Kokpunkt: 295 °C (1013 hPa)
Smältpunkt: 113…115 °C
Densitet: 1,203 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001111
CAS nummer: 86-73-7
EINECS: 201-695-5
UN: 3077
ADR: 9,III
Merck Index: 14,04155
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b4f703103bca335f93129e73cb008ee6ffa78a&vid=e5b4f703103bca335f93129e73cb008ee6ffa78a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 114.0 to 116.0 deg-C
Solubility in Toluene almost transparency