Dibensofuran ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Dibenzo[b,d]furan
D0147-25G D0147-500G D0147-100G
TCIAD0147-25GEA 430 SEK
TCIAD0147-25G TCIAD0147-500G TCIAD0147-100G
Dibensofuran ≥98.0%
Dibensofuran
Formel: C₁₂H₈O
MW: 168,19 g/mol
Kokpunkt: 285 °C (1013 hPa)
Smältpunkt: 79…82 °C
Densitet: 0,966 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00004968
CAS nummer: 132-64-9
EINECS: 205-071-3
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1227945463ecd35852e58ae10ef8786b0f71128&vid=d1227945463ecd35852e58ae10ef8786b0f71128&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in hot Methanol almost transparency