(±)-2-Methylpiperazine ≥99.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Methylpiperazine
M1179-25G M1179-500G
TCIAM1179-25GEA 295 SEK
TCIAM1179-25G TCIAM1179-500G
(±)-2-Methylpiperazine ≥99.0%
(±)-2-Methylpiperazine
Formel: C₅H₁₂N₂
MW: 100,16 g/mol
Kokpunkt: 153°C
Storage Temperature: Omgivande
MDL Number: MFCD00005954
CAS nummer: 109-07-9
EINECS: 203-644-2
UN: 2925
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b968a4bf8e57e259e3d739f5bdc242dc971978b8&vid=b968a4bf8e57e259e3d739f5bdc242dc971978b8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 63.0 to 67.0 deg-C