α-Terpinene ≥90.0%

Leverantör: TCI EUROPE
Fara

Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene

M0317-25ML M0317-500ML
TCIAM0317-25MLEA 215 SEK
TCIAM0317-25ML TCIAM0317-500ML
α-Terpinene ≥90.0%
α-Terpinene
Formel: C₁₀H₁₆
MW: 136,24 g/mol
Kokpunkt: 173°C
Smältpunkt: –48 °C
Densitet: 0,834 g/cm³ (20 °C)
Flampunkt: 52°C
Storage Temperature: Kylskåp
MDL Number: MFCD00001534
CAS nummer: 99-86-5
EINECS: 202-795-1
UN: 2319
ADR: 3,III
Merck Index: 14,09170
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f494ea3f3a3a8a64b3adc6da2cb4d2cbad20e1c8&vid=f494ea3f3a3a8a64b3adc6da2cb4d2cbad20e1c8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR