Indoline ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2,3-Dihydroindol

I0033-25ML I0033-250ML
TCIAI0033-25MLEA 285 SEK
TCIAI0033-25ML TCIAI0033-250ML
Indoline ≥98.0% (by GC, titration analysis)
Indoline
Formel: C₈H₉N
Kokpunkt: 231°C
Smältpunkt: –21 °C
Densitet: 1,0676 g/cm³ (20 °C)
Flampunkt: 93°C
Storage Temperature: Omgivande
MDL Number: MFCD00005705
CAS nummer: 496-15-1
EINECS: 207-816-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6326d06b43291a24e727d5d66bbedc1061fb6006&vid=6326d06b43291a24e727d5d66bbedc1061fb6006&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR