Perhydroazepine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Hexamethyleneimine
H0092-25ML H0092-500ML
TCIAH0092-25MLEA 215 SEK
TCIAH0092-25ML TCIAH0092-500ML
Perhydroazepine ≥98.0% (by GC, titration analysis)
Perhydroazepine
Formel: C₆H₁₃N
MW: 99,18 g/mol
Kokpunkt: 138°C
Smältpunkt: –37 °C
Densitet: 0,88 g/cm³ (20 °C)
Flampunkt: 18°C
Storage Temperature: Omgivande
MDL Number: MFCD00006934
CAS nummer: 111-49-9
EINECS: 203-875-9
UN: 2493
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=512bf354c9f162093351b0bb71dcfe41c2e0aad0&vid=512bf354c9f162093351b0bb71dcfe41c2e0aad0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %