cis-Stilbene ≥96.0%
Leverantör: TCI EUROPE
Varning
Synonyms: cis-1,2-Diphenylethene, Isostilbene, cis-1,2-Diphenylethylene, (Z)-1,2-Diphenylethene
S0089-5ML S0089-1ML
TCIAS0089-5MLEA 885 SEK
TCIAS0089-5ML TCIAS0089-1ML
cis-Stilbene ≥96.0%
cis-Stilbene
Formel: (C₆H₅)CHCH(C₆H₅)
MW: 180,25 g/mol
Kokpunkt: 152°C
Densitet: 1,01 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004788
CAS nummer: 645-49-8
EINECS: 211-445-7
Merck Index: 14,08817
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0a968ffbca7aced95ca6de5610cf523fd064c8d&vid=c0a968ffbca7aced95ca6de5610cf523fd064c8d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 96.0 %