Benzyl thiocyanate ≥99.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Thiocyanic acid Benzyl ester
T0198-25G
T0198-500G
TCIAT0198-25GEA
215
SEK
TCIAT0198-25G
TCIAT0198-500G
Benzyl thiocyanate ≥99.0%
Benzyl thiocyanate
Formel:
C₈H₇NS MW: 149.22 g/mol Kokpunkt: 256°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00001832 CAS nummer: 3012-37-1 EINECS: 221-144-2 UN: 1759 ADR: 8 II |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 40.0 to 43.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...