Kaliumdiväteortofosfat
Leverantör: G Biosciences
Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
RC-084 RC-083
RC-084EA 4025 SEK
RC-084 RC-083
Kaliumdiväteortofosfat
Kaliumdiväteortofosfat
Formel: KH₂PO₄
MW: 136.09 g/mol
Kokpunkt: ∼100 °C (1013 hPa)
Smältpunkt: 253
Densitet: 2.3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011401
CAS nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744