1,2,3-Butanetriol ≥90.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1,2,3-Trihydroxybutane, Butane-1,2,3-triol
B0918-25ML B0918-5ML
TCIAB0918-25MLEA 17175 SEK
TCIAB0918-25ML TCIAB0918-5ML
1,2,3-Butanetriol ≥90.0%
1,2,3-Butanetriol
Formel: C₄H₁₀O₃
MW: 106,12 g/mol
Kokpunkt: 175°C
MDL Number: MFCD00059662
CAS nummer: 4435-50-1
EINECS: 224-643-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ea9885bb3de121800a74fdd7233f1981929ebfc&vid=8ea9885bb3de121800a74fdd7233f1981929ebfc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %