2,5-Di-tert-butylhydroquinone ≥98.0%
Leverantör: TCI
Varning
Synonyms: BHQ
D0940-25G D0940-500G
TCIAD0940-25GEA 230 SEK
TCIAD0940-25G TCIAD0940-500G
2,5-Di-tert-butylhydroquinone ≥98.0%
2,5-Di-tert-butylhydroquinone
Formel: C₁₄H₂₂O₂
Kokpunkt: 334 °C (1013 hPa)
Smältpunkt: 212 °C
Densitet: 1,078 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008825
CAS nummer: 88-58-4
EINECS: 201-841-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3fad0297e4572a20354bb96a211949e410372c1&vid=a3fad0297e4572a20354bb96a211949e410372c1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 215.0 to 222.0 deg-C
Solubility in Methanol almost transparency