1,16-Hexadecanediol ≥95.0%

Leverantör: TCI EUROPE

Synonyms: Hexadecamethylene glycol, 1,16-Dihydroxyhexadecane

H0552-1G H0552-5G
TCIAH0552-1GEA 1450 SEK
TCIAH0552-1G TCIAH0552-5G
1,16-Hexadecanediol ≥95.0%
1,16-Hexadecanediol
Formel: C₁₆H₃₄O₂
MW: 258,44 g/mol
Kokpunkt: 200°C
MDL Number: MFCD00002821
CAS nummer: 7735-42-4
EINECS: 231-794-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=638b4b367e629c89d37ffd9e8ad928895eb2fc79&vid=638b4b367e629c89d37ffd9e8ad928895eb2fc79&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 90.0 to 94.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR