2-Hydroxypyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
H0330-25G H0330-250G
TCIAH0330-25GEA 605 SEK
TCIAH0330-25G TCIAH0330-250G
2-Hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Hydroxypyridine
Formel: C₅H₅NO
MW: 95,10 g/mol
Kokpunkt: 281°C
Smältpunkt: 107 °C
Densitet: 1,085 g/cm³ (30 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006268
CAS nummer: 142-08-5
EINECS: 205-520-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=121832894cc06a2488bae7682a1cbb964075997e&vid=121832894cc06a2488bae7682a1cbb964075997e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C