2-Allylphenol ≥98.0%

Leverantör: TCI
Fara

A0233-25G A0233-500G
TCIAA0233-25GEA 215 SEK
TCIAA0233-25G TCIAA0233-500G
2-Allylphenol ≥98.0%
2-Allylphenol
Formel: C₉H₁₀O
Kokpunkt: 220°C
Smältpunkt: –2 °C
Densitet: 1,03 g/cm³ (20 °C)
Flampunkt: 88°C
Storage Temperature: Omgivande
MDL Number: MFCD00002250
CAS nummer: 1745-81-9
EINECS: 217-119-0
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e55f2af9394b975ebf75085d725c8cbef89bf8b5&vid=e55f2af9394b975ebf75085d725c8cbef89bf8b5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR