(±)-3-Buten-2-ol ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
(±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol
B0695-25ML
B0695-5ML
TCIAB0695-25MLEA
2675
SEK
TCIAB0695-25ML
TCIAB0695-5ML
(±)-3-Buten-2-ol ≥97.0%
(±)-3-Buten-2-ol
Formel:
C₄H₈O MW: 72,11 g/mol Kokpunkt: 100°C Smältpunkt: –100 °C Densitet: 0,913 g/cm³ (20 °C) Flampunkt: 16°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004543 CAS nummer: 598-32-3 EINECS: 209-929-8 UN: 1986 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...