(R)-(-)-2-Butanol ≥99.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol
B0926-1ML B0926-5ML
TCIAB0926-1MLEA 645 SEK
TCIAB0926-1ML TCIAB0926-5ML
(R)-(-)-2-Butanol ≥99.0%
(R)-(-)-2-Butanol
Formel: C₄H₁₀O
MW: 74,12 g/mol
Kokpunkt: 100°C
Smältpunkt: –114 °C
Densitet: 0,807 g/cm³ (20 °C)
Flampunkt: 24°C
Storage Temperature: Omgivande
MDL Number: MFCD00064280
CAS nummer: 14898-79-4
EINECS: 238-967-8
UN: 1120
ADR: 3,III
Merck Index: 14,01541
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75068adc0e890b6206e744ac10dac7fe5ae9842c&vid=75068adc0e890b6206e744ac10dac7fe5ae9842c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Specific rotation [a]20/D -11.0 to -14.0 deg (neat)