Cyclobutylméthanol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Cyclobutanemethanol, Cyclobutylmethanol, Hydroxymethylcyclobutane

C2061-25G C2061-5G
TCIAC2061-25GEA 3050 SEK
TCIAC2061-25G TCIAC2061-5G
Cyclobutylméthanol ≥98.0%
Cyclobutylméthanol
Formel: C₅H₁₀O
MW: 86,13 g/mol
Kokpunkt: 144°C
Densitet: 0,92 g/cm³ (20 °C)
Flampunkt: 40°C
Storage Temperature: Omgivande
MDL Number: MFCD00001330
CAS nummer: 4415-82-1
EINECS: 224-575-4
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f64bcbd70087f2d4b6d77bc114f7d6f67b955629&vid=f64bcbd70087f2d4b6d77bc114f7d6f67b955629&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR