(3-Mercaptopropyl)triethoxysilane ≥96.0%
Leverantör: TCI
Varning
M1505-25G M1505-500G M1505-100G
TCIAM1505-25GEA 335 SEK
TCIAM1505-25G TCIAM1505-500G TCIAM1505-100G
(3-Mercaptopropyl)triethoxysilane ≥96.0%
(3-Mercaptopropyl)triethoxysilane
Formel: HSH₂CCH₂CH₂Si(OCH₂CH₃)₃
Kokpunkt: 115°C
Flampunkt: 88°C
Storage Temperature: Kylskåp
MDL Number: MFCD00053924
CAS nummer: 14814-09-6
EINECS: 238-883-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11fa23e48013f81b4cbf283b4f4bd0cf07d75b92&vid=11fa23e48013f81b4cbf283b4f4bd0cf07d75b92&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %