Phenyltriethoxysilane ≥99.0%
Leverantör: TCI
|
Varning
|
P0735-25ML
P0735-500ML
P0735-100ML
TCIAP0735-25MLEA
215
SEK
TCIAP0735-25ML
TCIAP0735-500ML
TCIAP0735-100ML
Phenyltriethoxysilane ≥99.0%
Phenyltriethoxysilane
Formel:
C₁₂H₂₀O₃Si Kokpunkt: 235°C Smältpunkt: 41 °C Densitet: 0,996 g/cm³ (20 °C) Flampunkt: 111°C |
MDL Number:
MFCD00009065 CAS nummer: 780-69-8 EINECS: 212-305-8 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...