Vinyltriethoxysilane ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Triethoxy(vinyl)silane
V0044-25ML
V0044-500ML
V0044-100ML
TCIAV0044-25MLEA
215
SEK
TCIAV0044-25ML
TCIAV0044-500ML
TCIAV0044-100ML
Vinyltriethoxysilane ≥98.0%
Vinyltriethoxysilane
Formel:
C₈H₁₈O₃Si Kokpunkt: 161°C Densitet: 0,91 g/cm³ (20 °C) Flampunkt: 34°C Storage Temperature: Omgivande |
MDL Number:
MFCD00009063 CAS nummer: 78-08-0 EINECS: 201-081-7 UN: 1993 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...