2-Methoxythiophenol ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-Methoxybenzenethiol, o-Methoxythiophenol, 2-Mercaptoanisol, 2-Methoxyphenyl mercaptan, 2-Methoxybenzene-1-thiol, 1-Methoxy-2-mercaptobenzene

M1670-25G M1670-5G
TCIAM1670-25GEA 2850 SEK
TCIAM1670-25G TCIAM1670-5G
2-Methoxythiophenol ≥98.0%
2-Methoxythiophenol
Formel: C₇H₈OS
Kokpunkt: 219°C
Densitet: 1,14 g/cm³ (30 °C)
Flampunkt: 95°C
MDL Number: MFCD00004834
CAS nummer: 7217-59-6
EINECS: 230-605-7
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6117229ff02b79424e7d535ad6bccc139c96c4b&vid=c6117229ff02b79424e7d535ad6bccc139c96c4b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR