4-Allylanisole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1-Allyl-4-methoxybenzene, Estragole

A0702-10G A0702-100G
TCIAA0702-10GEA 455 SEK
TCIAA0702-10G TCIAA0702-100G
4-Allylanisole ≥98.0%
4-Allylanisole
Formel: C₁₀H₁₂O
MW: 148.2 g/mol
Kokpunkt: 220°C
Smältpunkt: 22.5
Flampunkt: 81°C
MDL Number: MFCD00008653
CAS nummer: 140-67-0
EINECS: 205-427-8
Merck Index: 14,03705
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fbd13ef26bb83b14fdaafbe5bdb1cb25567e47d&vid=6fbd13ef26bb83b14fdaafbe5bdb1cb25567e47d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR