2-Nitrofluorene ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Nitro-9H-fluorene
N0201-25G
N0201-100G
TCIAN0201-25GEA
655
SEK
TCIAN0201-25G
TCIAN0201-100G
2-Nitrofluorene ≥99.0%
2-Nitrofluorene
Formel:
C₁₃H₉NO₂ MW: 211,22 g/mol Smältpunkt: 154…157 °C |
MDL Number:
MFCD00001117 CAS nummer: 607-57-8 EINECS: 210-138-5 UN: 3077 ADR: 9,III |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Melting point | 155.0 to 159.0 deg-C |
Solubility in hot Acetic acid | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...