2,4,6-Triaminopyrimidine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Pyrimidine-2,4,6-triyltriamine

T0834-25G T0834-500G
TCIAT0834-25GEA 365 SEK
TCIAT0834-25G TCIAT0834-500G
2,4,6-Triaminopyrimidine ≥98.0% (by HPLC, titration analysis)
2,4,6-Triaminopyrimidine
Formel: C₄H₇N₅
MW: 125,13 g/mol
Kokpunkt: 324 °C (1013 hPa)
Smältpunkt: 249°C
Densitet: 1,45 g/cm³ (20 °C)
MDL Number: MFCD00006100
CAS nummer: 1004-38-2
EINECS: 213-720-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27026141779d8a252b42a0c8e96f58e5ec6fa2b0&vid=27026141779d8a252b42a0c8e96f58e5ec6fa2b0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR