4,4'-Metylendianilin ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
4,4'-Diaminodifenylmetan
M0220-500G
M0220-25G
TCIAM0220-500GEA
495
SEK
TCIAM0220-500G
TCIAM0220-25G
4,4'-Metylendianilin ≥98.0%
4,4'-Metylendianilin
Formel:
C₁₃H₁₄N₂ MW: 198,27 g/mol Kokpunkt: 257°C Smältpunkt: 91…92 °C Densitet: 1,15 g/cm³ (20 °C) Flampunkt: 220 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00007914 CAS nummer: 101-77-9 EINECS: 202-974-4 UN: 2651 ADR: 6.1,III Merck Index: 14,02980 |
Specifikation testresultat
Appearance | White to Orange to Green pellet |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 91.0 to 94.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...