o-Fenylendiamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 1,2-Fenylendiamin
P0168-500G P0168-25G P0168-100G
TCIAP0168-500GEA 2000 SEK
TCIAP0168-500G TCIAP0168-25G TCIAP0168-100G
o-Fenylendiamin ≥98.0% (by GC, titration analysis)
o-Fenylendiamin
Formel: C₆H₈N₂
MW: 108,14 g/mol
Kokpunkt: 265°C
Smältpunkt: 103 °C
Densitet: 1,21 g/cm³ (20 °C)
Flampunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007721
CAS nummer: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1,III
Merck Index: 14,07284
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eec6a0126029e74a3a232b4fbde4bd14a20588f1&vid=eec6a0126029e74a3a232b4fbde4bd14a20588f1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 100.0 to 105.0 deg-C