4-Aminoindole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Indol-4-ylamine, 4-Indolamine, 4-Amino-1H-indole

A2070-1G A2070-5G
TCIAA2070-1GEA 1525 SEK
TCIAA2070-1G TCIAA2070-5G
4-Aminoindole ≥98.0%
4-Aminoindole
Formel: C₈H₈N₂
MW: 132,16 g/mol
Smältpunkt: 108 °C
MDL Number: MFCD01076559
CAS nummer: 5192-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb13fa88b79f15cb9a1f2dba83a7c6d01b85285c&vid=bb13fa88b79f15cb9a1f2dba83a7c6d01b85285c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR