Benzyl isothiocyanate ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Benzylisothiocyanate
I0224-25G
I0224-5G
TCIAI0224-25GEA
1325
SEK
TCIAI0224-25G
TCIAI0224-5G
Benzyl isothiocyanate ≥98.0%
Benzyl isothiocyanate
Formel:
C₈H₇NS MW: 149.22 g/mol Kokpunkt: 243°C Smältpunkt: 24 Storage Temperature: Kylskåp |
MDL Number:
MFCD00004819 CAS nummer: 622-78-6 EINECS: 210-753-9 UN: 3082 ADR: 9 III |
|
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...