Isobutyl isothiocyanate ≥97.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Methylpropyl isothiocyanate
I0186-25ML I0186-5ML
TCIAI0186-25MLEA 925 SEK
TCIAI0186-25ML TCIAI0186-5ML
Isobutyl isothiocyanate ≥97.0%
Isobutyl isothiocyanate
Formel: C₅H₉NS
MW: 115,2 g/mol
Kokpunkt: 164°C
Densitet: 0,94 g/cm³ (20 °C)
Flampunkt: 24°C
Storage Temperature: Omgivande
MDL Number: MFCD00022062
CAS nummer: 591-82-2
EINECS: 209-733-2
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cad6cefd910871c3524d43b176257d20958f4d2&vid=1cad6cefd910871c3524d43b176257d20958f4d2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %