N,N-Dietyl-m-toluidin ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 3-Diethylaminotoluene, N,N-Diethyl-3-methylaniline, DMT
D0532-25ML D0532-500ML
TCIAD0532-25MLEA 285 SEK
TCIAD0532-25ML TCIAD0532-500ML
N,N-Dietyl-m-toluidin ≥99.0% (by GC, titration analysis)
N,N-Dietyl-m-toluidin
Formel: C₁₁H₁₇N
MW: 163,26 g/mol
Kokpunkt: 107°C
Densitet: 0,9051 g/cm³ (15 °C)
Flampunkt: 104°C
MDL Number: MFCD00035795
CAS nummer: 91-67-8
EINECS: 202-089-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04b046e6d811ffbcbd810fcde497659b84712cc9&vid=04b046e6d811ffbcbd810fcde497659b84712cc9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %