5-Iodo-2-furaldehyde ≥98.0%

Leverantör: TCI
Varning

Synonyms: 5-Iodofurfural

I0696-1G I0696-5G
TCIAI0696-1GEA 485 SEK
TCIAI0696-1G TCIAI0696-5G
5-Iodo-2-furaldehyde ≥98.0%
5-Iodo-2-furaldehyde
Formel: C₅H₃IO₂
MW: 221.98 g/mol
MDL Number: MFCD00159503
CAS nummer: 2689-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=659cc52d9e26de515ebc0f92eefcbfc794474369&vid=659cc52d9e26de515ebc0f92eefcbfc794474369&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR