Benzyltriethylammonium iodide ≥98.0%

Leverantör: TCI
Varning

B0446-25G B0446-5G
TCIAB0446-25GEA 1400 SEK
TCIAB0446-25G TCIAB0446-5G
Benzyltriethylammonium iodide ≥98.0%
Benzyltriethylammonium iodide
Formel: C₁₃H₂₂IN
MDL Number: MFCD00043178
CAS nummer: 5400-94-2
EINECS: 226-446-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e29775a4496995a475db98666d4c5076981e0b1d&vid=e29775a4496995a475db98666d4c5076981e0b1d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 168.0 to 173.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR