Guanidiniumtiocyanat ≥99.0%
Leverantör: TCI
Varning
Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
G0230-25G G0230-500G
TCIAG0230-25GEA 485 SEK
TCIAG0230-25G TCIAG0230-500G
Guanidiniumtiocyanat ≥99.0%
Guanidiniumtiocyanat
Formel: CH₅N₃·CHNS
MW: 118.16 g/mol
Smältpunkt: 118
Storage Temperature: Omgivande
MDL Number: MFCD00013027
CAS nummer: 593-84-0
EINECS: 209-812-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cd35aac3ceef11179009094d87c8aec0b08785d&vid=1cd35aac3ceef11179009094d87c8aec0b08785d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 118.0 to 122.0 deg-C
Solubility in Water almost transparency