2,3,4,5,6-Pentabromobenzyl alcohol ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
(2,3,4,5,6-Pentabromophenyl)methanol, Pentabromobenzyl alcohol, 2,3,4,5,6-Pentabromobenzylalcohol
P1457-1G
P1457-5G
TCIAP1457-1GEA
325
SEK
TCIAP1457-1G
TCIAP1457-5G
2,3,4,5,6-Pentabromobenzyl alcohol ≥98.0%
2,3,4,5,6-Pentabromobenzyl alcohol
Formel:
C₇H₃Br₅O Kokpunkt: 395 °C (1013 hPa) Smältpunkt: 272 °C |
MDL Number:
MFCD00192377 CAS nummer: 79415-41-1 |
|
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...