2-Fluoro-4-nitroanisole ≥98.0%
Leverantör: TCI
Varning
F0617-1G F0617-5G
TCIAF0617-1GEA 215 SEK
TCIAF0617-1G TCIAF0617-5G
2-Fluoro-4-nitroanisole ≥98.0%
2-Fluoro-4-nitroanisole
Formel: C₇H₆FNO₃
Storage Temperature: Omgivande
MDL Number: MFCD00061095
CAS nummer: 455-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500a2e3628eba12fa7368dba8d1ee4b04735e29d&vid=500a2e3628eba12fa7368dba8d1ee4b04735e29d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 107.0 deg-C