α,α,α-Trifluoroacetophenone ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone
T0848-25G T0848-5G
TCIAT0848-25GEA 1475 SEK
TCIAT0848-25G TCIAT0848-5G
α,α,α-Trifluoroacetophenone ≥98.0%
α,α,α-Trifluoroacetophenone
Formel: C₈H₅F₃O
MW: 174,12 g/mol
Kokpunkt: 166°C
Smältpunkt: –40 °C
Densitet: 1,2799 g/cm³ (20 °C)
Flampunkt: 41°C
Storage Temperature: Omgivande
MDL Number: MFCD00000420
CAS nummer: 434-45-7
EINECS: 207-103-1
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e57914d51cd97795b56ff0a06e00d4e204f7165&vid=1e57914d51cd97795b56ff0a06e00d4e204f7165&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %